Thermo Scientific Chemicals

2-Hydroxy-6-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 156390050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxy-6-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 156390050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156390050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3279-76-3
IUPAC Name
6-methyl-1,2-dihydropyridin-2-one
Molecular Formula
C6H7NO
InChI Key
JEAVIRYCMBDJIU-UHFFFAOYSA-N
SMILES
CC1=CC=CC(=O)N1
Appearance (Color)
White to light brown
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Form)
Crystalline powder
Melting point
155°C to 160°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text