Penicillin-Streptomycin-Neomycin (PSN) Antibiotic Mixture
Save up to 29% on your order of cell culture products. View promotion details >
Product Image
Gibco™

Penicillin-Streptomycin-Neomycin (PSN) Antibiotic Mixture

The antibiotics Penicillin, Streptomycin, and Neomycin are used to prevent bacterial contamination of cell cultures due to their effective combinedRead more
Promo ImagePromotion
Promo code:RCELL25
Save up to 29% across the cell culture workflow - research begins with cells
From now until Dec 31, get up to 29% off media, sera, buffers, reagents and other cell culture products to advance your researchLearn More
Have Questions?
Catalog NumberQuantity
15640055Promo Image100 mL
Catalog number 15640055
Price (USD)
51.65
Online Exclusive
53.50
Save 1.85 (3%)
Each
In stock
Add to cart
Quantity:
100 mL
Price (USD)
51.65
Online Exclusive
53.50
Save 1.85 (3%)
Each
Add to cart

The antibiotics Penicillin, Streptomycin, and Neomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Penicillin was originally purified from the fungus Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus and Neomycin from Streptomyces fradiae. Streptomycin and Neomycin act by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. We offer a variety of antibiotics and antimycotics for cell culture applications.

This solution contains 5 mg of penicillin, 5 mg of streptomycin, and 10 mg of neomycin per ml in 0.85% saline.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration100 X
Culture TypeMammalian Cell Culture
For Use With (Application)Prevention of Cell Culture Contamination
Quantity100 mL
Shipping ConditionDry Ice
FormLiquid
Product TypeAntibiotic
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -20°C).

Frequently asked questions (FAQs)

How can I decontaminate my cultures?

When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.

1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.

The following is a suggested procedure for determining toxicity levels and decontaminating cultures:

1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What antibiotics do you offer to help control or eliminate cell culture contamination?

Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link