6-Hydroxynicotinic acid, 99%, Thermo Scientific Chemicals
6-Hydroxynicotinic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Hydroxynicotinic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 156401000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5006-66-6
IUPAC Name6-oxo-1,6-dihydropyridine-3-carboxylic acid
Molecular FormulaC6H5NO3
InChI KeyBLHCMGRVFXRYRN-UHFFFAOYSA-N
SMILESOC(=O)C1=CNC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH98.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only