3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals

3,5-dinitrosalicylic acid, CAS # 609-99-4, also known as DNS or DNSA, is used for quantifying carbohydrate levels in blood by calorimetry.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 156440010
Price (USD)/ Each
493.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
581.00 
Save 87.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
493.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
581.00 
Save 87.35 (15%)
Add to cart
3,5-Dinitrosalicylic acid, 98%, Thermo Scientific Chemicals
Catalog number156440010
Price (USD)/ Each
493.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
581.00 
Save 87.35 (15%)
-
Add to cart
Chemical Identifiers
CAS609-99-4
IUPAC Name2-hydroxy-3,5-dinitrobenzoic acid
Molecular FormulaC7H4N2O7
InChI KeyLWFUFLREGJMOIZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point170°C to 174°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

3,5-Dinitrosalicylic acid also known as DNA or DNSA reacts with reducing sugars forming 3-amino-5-nitrosalicylic acid, which is an aromatic compound with maximum absorption at 540 nm allowing spectrophotometer measurement of the amount of reducing molecules

Applications

  • It is used in the quantification of reducing sugars by colorimetry in the blood. It is also used to analyze glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar
  • 3,5-Dinitrosalicylic acid can detect reducing substances in the urine
  • This compound is used to detect the presence of free carbonyl groups (C=O)
  • 3,5-Dinitrosalicylic acid can be used to determine the effects of silver nanoparticles (AgNPs) on membrane leakage
RUO – Research Use Only