Tribenzylamine, 99+%, Thermo Scientific Chemicals
Tribenzylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tribenzylamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
Catalog number 156470010
Price (USD)/ Each
283.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
283.00
Add to cart
Tribenzylamine, 99+%, Thermo Scientific Chemicals
Catalog number156470010
Price (USD)/ Each
283.00
-
Add to cart
Chemical Identifiers
IUPAC Nametribenzylamine
Molecular FormulaC21H21N
InChI KeyMXHTZQSKTCCMFG-UHFFFAOYSA-N
SMILESC(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)287.41
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
GC>=99.0 %
Infrared spectrumConforms
Titration with HClO4>=99.0 % (On anhydrous substance)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only