Thermo Scientific Chemicals

Tribenzylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 156471000
100 g, Each
Thermo Scientific Chemicals

Tribenzylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 156471000
100 g, Each
Quantity
Catalog number: 156471000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-40-6
IUPAC Name
tribenzylamine
Molecular Formula
C21H21N
InChI Key
MXHTZQSKTCCMFG-UHFFFAOYSA-N
SMILES
C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream
GC
>=99.0 %
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 % (On anhydrous substance)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text