Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKAPGRLVLI ILCSVVFSAV YILLCCWAGL PLCLATCLDH HFPTGSRPTV PGPLHFSGYS SVPDGKPLVR EPCRSCAVVS SSGQMLGSGL GAEIDSAECV FRMNQAPTVG FEADVGQRST LRVVSHTSVP LLLRNYSHYF QKARDTLYMV WGQGRHMDRV LGGRTYRTLL QLTRMYPGLQ VYTFTERMMA YCDQIFQDET GKNRRQSGSF LSTGWFTMIL ALELCEEIVV YGMVSDSYCR EKSHPSVPYH YFEKGRLDEC QMYLAHEQAP RSAHRFITEK AVFSRWAKKR PIVFAHPSWR TE
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein prefers glycoproteins rather than glycolipids as substrates and shows restricted substrate specificity, utilizing only the trisaccharide sequence Neu5Ac-alpha-2,3-Gal-beta-1,3-GalNAc. In addition, it is involved in the synthesis of ganglioside GD1A from GM1B. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-N-acetyl-galactos; Alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3-N-acetyl-galactosaminide alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2, 6-sialyltransferase alpha2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase IV; Sialyltransferase 3C; Sialyltransferase 7D; sialyltransferase 7D ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase); SIAT3-C; SIAT7-D; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 GalNAc alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 4; ST6GalNAc IV; ST6GalNAcIV
Gene Aliases: IV; SIAT3-C; SIAT3C; SIAT7-D; SIAT7D; ST6GalNAc; ST6GALNAC4; ST6GALNACIV
UniProt ID: (Human) Q9H4F1
Entrez Gene ID: (Human) 27090
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support