Thermo Scientific Chemicals

Ethyl 5-bromovalerate, 99%, Thermo Scientific Chemicals

Catalog number: 156541000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 5-bromovalerate, 99%, Thermo Scientific Chemicals

Catalog number: 156541000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 156541000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14660-52-7
IUPAC Name
ethyl 5-bromopentanoate
Molecular Formula
C7H13BrO2
InChI Key
AFRWBGJRWRHQOV-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCCBr
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4570 to 1.4600 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text