2,3-Dimethyl-2-butene, 98%
2,3-Dimethyl-2-butene, 98%
2,3-Dimethyl-2-butene, 98%
2,3-Dimethyl-2-butene, 98%
Thermo Scientific Chemicals

2,3-Dimethyl-2-butene, 98%

CAS: 563-79-1 | C6H12 | 84.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
25 mL
Packaging:
Glass bottle
Catalog number 156590250
Price (USD)
75.65
Special offer
Online exclusive
Ends: 26-Sep-2025
93.50
Save 17.85 (19%)
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 26-Sep-2025
93.50
Save 17.85 (19%)
Each
Add to cart
Chemical Identifiers
CAS563-79-1
IUPAC Name2,3-dimethylbut-2-ene
Molecular FormulaC6H12
InChI KeyWGLLSSPDPJPLOR-UHFFFAOYSA-N
SMILESCC(C)=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4110 to 1.4130 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection