4-Iodoaniline, 99%, Thermo Scientific Chemicals
4-Iodoaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodoaniline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 156611000
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
146.00 
Save 21.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
146.00 
Save 21.35 (15%)
Add to cart
4-Iodoaniline, 99%, Thermo Scientific Chemicals
Catalog number156611000
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
146.00 
Save 21.35 (15%)
-
Add to cart
Chemical Identifiers
CAS540-37-4
IUPAC Name4-iodoaniline
Molecular FormulaC6H6IN
InChI KeyVLVCDUSVTXIWGW-UHFFFAOYSA-N
SMILESNC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to grey-brown to dark grey
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point61°C to 64°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only