5-Iodovanillin, 97%, Thermo Scientific Chemicals
5-Iodovanillin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Iodovanillin, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 156670250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-hydroxy-3-iodo-5-methoxybenzaldehyde
Molecular FormulaC8H7IO3
InChI KeyFBBCSYADXYILEH-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC(I)=C1O
Molecular Weight (g/mol)278.05
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point180°C to 184°C
HPLC>=96.0 %
Appearance (Color)Light yellow
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only