Thermo Scientific Chemicals

Isovaleric acid, 99%, Thermo Scientific Chemicals

Catalog number: 156691000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Isovaleric acid, 99%, Thermo Scientific Chemicals

Catalog number: 156691000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156691000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
503-74-2
IUPAC Name
3-methylbutanoic acid
Molecular Formula
C5H10O2
InChI Key
GWYFCOCPABKNJV-UHFFFAOYSA-N
SMILES
CC(C)CC(O)=O
Refractive index
1.3990 to 1.4070 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text