Thermo Scientific Chemicals

4-(2-Keto-1-benzimidazolinyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: 156700250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(2-Keto-1-benzimidazolinyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: 156700250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156700250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20662-53-7
IUPAC Name
1-(piperidin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular Formula
C12H15N3O
InChI Key
BYNBAMHAURJNTR-UHFFFAOYSA-N
SMILES
O=C1NC2=CC=CC=C2N1C1CCNCC1
Appearance (Form)
Powder
Appearance (Color)
Off-white to light yellow
Infrared spectrum
Conforms
Melting point
183°C to 185°C
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text