Thermo Scientific Chemicals

Maleimide, 98%, Thermo Scientific Chemicals

Catalog number: 156715000
500 g, Glass Bottle, Each
Thermo Scientific Chemicals

Maleimide, 98%, Thermo Scientific Chemicals

Catalog number: 156715000
500 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 156715000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
541-59-3
IUPAC Name
potassium silver(1+) bis(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-ide)
Molecular Formula
C8H4AgKN2O4
InChI Key
KWLWLQRUFFGWNG-UHFFFAOYSA-L
SMILES
[K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1
Infrared spectrum
Conforms
Melting point
90°C to 94°C
Titration with NaOH
>=97.5 %
GC
>=97.5 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text