Thermo Scientific Chemicals

Malonaldehyde bis(diethyl acetal), 97%, Thermo Scientific Chemicals

Catalog number: 156731000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Malonaldehyde bis(diethyl acetal), 97%, Thermo Scientific Chemicals

Catalog number: 156731000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 156731000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
122-31-6
IUPAC Name
1,1,3,3-tetraethoxypropane
Molecular Formula
C11H24O4
InChI Key
KVJHGPAAOUGYJX-UHFFFAOYSA-N
SMILES
CCOC(CC(OCC)OCC)OCC
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4100 to 1.4120 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text