Malonaldehyde bis(diethyl acetal), 97%, Thermo Scientific Chemicals
Malonaldehyde bis(diethyl acetal), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Malonaldehyde bis(diethyl acetal), 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number 156735000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-31-6
IUPAC Name1,1,3,3-tetraethoxypropane
Molecular FormulaC11H24O4
InChI KeyKVJHGPAAOUGYJX-UHFFFAOYSA-N
SMILESCCOC(CC(OCC)OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only