Thermo Scientific Chemicals

2-Mercaptopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 156740100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Mercaptopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 156740100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156740100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1450-85-7
IUPAC Name
1,2-dihydropyrimidine-2-thione
Molecular Formula
C4H4N2S
InChI Key
HBCQSNAFLVXVAY-UHFFFAOYSA-N
SMILES
S=C1NC=CC=N1
Appearance (Color)
Yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text