Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals
Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 156790050
Price (USD)/ Each
15.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
18.20 
Save 2.55 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
15.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
18.20 
Save 2.55 (14%)
Add to cart
Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals
Catalog number156790050
Price (USD)/ Each
15.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
18.20 
Save 2.55 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethyl (2E)-3-aminobut-2-enoate
Molecular FormulaC5H9NO2
InChI KeyXKORCTIIRYKLLG-ONEGZZNKSA-N
SMILESCOC(=O)C=C(/C)N
Molecular Weight (g/mol)115.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point81°C to 85°C
Appearance (Color)White to yellow
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only