Thermo Scientific Chemicals

Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals

Catalog number: 156791000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals

Catalog number: 156791000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 156791000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14205-39-1
IUPAC Name
methyl (2E)-3-aminobut-2-enoate
Molecular Formula
C5H9NO2
InChI Key
XKORCTIIRYKLLG-ONEGZZNKSA-N
SMILES
COC(=O)C=C(/C)N
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
81°C to 85°C
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text