4-Methylaminophenol sulfate, 99%, extra pure, Thermo Scientific Chemicals
4-Methylaminophenol sulfate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylaminophenol sulfate, 99%, extra pure, Thermo Scientific Chemicals

4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Catalog number 156800025
Price (USD)/ Each
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
IUPAC Namebis(4-(methylamino)phenol); sulfuric acid
Molecular FormulaC14H20N2O6S
InChI KeyZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
Molecular Weight (g/mol)344.38
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<30 ppm
Residue after ignition=<0.15 %
Iron (Fe)=<50 ppm
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to beige to grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only