Thermo Scientific Chemicals

4-Methylaminophenol sulfate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 156805000
500 g, Each
Thermo Scientific Chemicals

4-Methylaminophenol sulfate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 156805000
500 g, Each
Quantity
Catalog number: 156805000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
55-55-0
IUPAC Name
bis(4-(methylamino)phenol); sulfuric acid
Molecular Formula
C14H20N2O6S
InChI Key
ZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
Appearance (Color)
White to beige to grey
Heavy metals (as Pb)
=<30 ppm
Residue after ignition
=<0.15 %
Iron (Fe)
=<50 ppm
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text