Methylcyclopentadiene dimer, 90%, stabilized, Thermo Scientific Chemicals
Methylcyclopentadiene dimer, 90%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylcyclopentadiene dimer, 90%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Glass bottle
Catalog number 156850025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26472-00-4
IUPAC Name4,9-dimethyltricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular FormulaC12H16
InChI KeyIYQYZZHQSZMZIG-UHFFFAOYNA-N
SMILESCC1=CC2C(C1)C1CC2C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4940 to 1.5020 (20°C, 589 nm)
GC>=88 % (Sum of isomers)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only