Thermo Scientific Chemicals

Methyl 2-furoate, 98%, Thermo Scientific Chemicals

Catalog number: 156870250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 2-furoate, 98%, Thermo Scientific Chemicals

Catalog number: 156870250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156870250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
611-13-2
IUPAC Name
methyl furan-2-carboxylate
Molecular Formula
C6H6O3
InChI Key
HDJLSECJEQSPKW-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CO1
Refractive index
1.4830 to 1.4890 (20°C, 589 nm)
Appearance (Color)
Clear light yellow to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text