Thermo Scientific Chemicals

Methyl methanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 156891000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl methanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 156891000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 156891000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
66-27-3
IUPAC Name
methyl methanesulfonate
Molecular Formula
C2H6O3S
InChI Key
MBABOKRGFJTBAE-UHFFFAOYSA-N
SMILES
COS(C)(=O)=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4130 to 1.4150 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkyl transfer reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text