1-Methylpiperazine, 99%, Thermo Scientific Chemicals
1-Methylpiperazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methylpiperazine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 156911000
Price (USD)/ Each
36.65
Online exclusive
42.70 
Save 6.05 (14%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
36.65
Online exclusive
42.70 
Save 6.05 (14%)
Add to cart
1-Methylpiperazine, 99%, Thermo Scientific Chemicals
Catalog number156911000
Price (USD)/ Each
36.65
Online exclusive
42.70 
Save 6.05 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-methylpiperazine
Molecular FormulaC5H12N2
InChI KeyPVOAHINGSUIXLS-UHFFFAOYSA-N
SMILESCN1CCNCC1
Molecular Weight (g/mol)100.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.4645 to 1.4665 (20°C, 589 nm)
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only