Thermo Scientific Chemicals

Methyltriphenylphosphonium Bromide, 98%, Thermo Scientific Chemicals

Catalog number: 156955000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Methyltriphenylphosphonium Bromide, 98%, Thermo Scientific Chemicals

Catalog number: 156955000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 156955000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1779-49-3
IUPAC Name
methyltriphenylphosphanium bromide
Molecular Formula
C19H18BrP
InChI Key
LSEFCHWGJNHZNT-UHFFFAOYSA-M
SMILES
[Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 %
Appearance (Form)
Powder
Melting point
230°C to 234°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text