4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 157051000
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
Add to cart
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Catalog number157051000
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
-
Add to cart
Chemical Identifiers
CAS100-02-7
IUPAC Name4-nitrophenol
Molecular FormulaC6H5NO3
InChI KeyBTJIUGUIPKRLHP-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point112°C to 115°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only