Gentamicin (10 mg/mL)
Save up to 29% on your order of cell culture products. View promotion details >
Gentamicin (10 mg/mL)
Gibco™

Gentamicin (10 mg/mL)

Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1571007210 x 10 mL
1571006410 mL
Catalog number 15710072
Price (USD)
166.65
Online Exclusive
178.00
Save 11.35 (6%)
Each
-
Add to cart
Quantity:
10 x 10 mL
Price (USD)
166.65
Online Exclusive
178.00
Save 11.35 (6%)
Each
Add to cart
Ask our AI about this Product
Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria. Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination on cell cultures. The recommended working concentration ranges from 0.5 to 50 μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Dual-Site cGMP Manufacturing
Gibco™ Gentamicin is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer a comparable Gibco™ Gentamicin product made in our Scotland facility (15710-049). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration10 mg/mL
Culture TypeMammalian Cell Culture
For Use With (Application)Prevention of Cell Culture Contamination
Quantity10 x 10 mL
Shelf Life24 Months
Shipping ConditionRoom Temperature
FormLiquid
Product TypeAntibiotic
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Storage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link