2-Furoic acid hydrazide, 98%, Thermo Scientific Chemicals
2-Furoic acid hydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Furoic acid hydrazide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 157160050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namefuran-2-carbohydrazide
Molecular FormulaC5H6N2O2
InChI KeySKTSVWWOAIAIKI-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CO1
Molecular Weight (g/mol)126.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Melting point76°C to 80°C
Appearance (Color)Beige
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only