Thermo Scientific Chemicals

Tris(2-butoxyethyl) phosphate, 95%, Thermo Scientific Chemicals

Catalog number: 157241000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(2-butoxyethyl) phosphate, 95%, Thermo Scientific Chemicals

Catalog number: 157241000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 157241000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
78-51-3
IUPAC Name
tris(2-butoxyethyl) phosphate
Molecular Formula
C18H39O7P
InChI Key
WTLBZVNBAKMVDP-UHFFFAOYSA-N
SMILES
CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
GC
>=94 %
Infrared spectrum
Conforms
Refractive index
1.4360 to 1.4390 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text