Thermo Scientific Chemicals

N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals

Catalog number: 157271000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals

Catalog number: 157271000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 157271000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6066-82-6
IUPAC Name
1-hydroxypyrrolidine-2,5-dione
Molecular Formula
C4H5NO3
InChI Key
NQTADLQHYWFPDB-UHFFFAOYSA-N
SMILES
ON1C(=O)CCC1=O
Solubility
(5 g/ 100 ml Acetone) Clear to light opalescent solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text