DL-4-Chlorophenylalanine, 98%
DL-4-Chlorophenylalanine, 98%
Thermo Scientific Chemicals

DL-4-Chlorophenylalanine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number 157280050
Price (USD)/ Each
103.65
Online exclusive
122.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
103.65
Online exclusive
122.00 
Save 18.35 (15%)
Add to cart
DL-4-Chlorophenylalanine, 98%
Catalog number157280050
Price (USD)/ Each
103.65
Online exclusive
122.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-amino-3-(4-chlorophenyl)propanoic acid
Molecular FormulaC9H10ClNO2
InChI KeyNIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILESNC(CC1=CC=C(Cl)C=C1)C(O)=O
Molecular Weight (g/mol)199.63
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only