Thermo Scientific Chemicals

Uracil, 99+%

Catalog number: 157300250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Uracil, 99+%

Catalog number: 157300250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 157300250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66-22-8
IUPAC Name
1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C4H4N2O2
InChI Key
ISAKRJDGNUQOIC-UHFFFAOYSA-N
SMILES
O=C1NC=CC(=O)N1
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=99.0 %
Sulfated ash
=<0.3 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text