Chemical Identifiers
CAS860-22-0
IUPAC Namedisodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
Molecular FormulaC16H8N2Na2O8S2
InChI KeyKHLVKKOJDHCJMG-QDBORUFSSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
View more