Indigo Carmine, High Purity Biological Stain
Indigo Carmine, High Purity Biological Stain
Thermo Scientific Chemicals

Indigo Carmine, High Purity Biological Stain

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 157445000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS860-22-0
IUPAC Namedisodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
Molecular FormulaC16H8N2Na2O8S2
InChI KeyKHLVKKOJDHCJMG-QDBORUFSSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to purple
Dye content>=80 % (On dried basis)
Insoluble matter=<0.2 % (in water)
Cadmium (Cd)=<1 ppm
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only