Thermo Scientific Chemicals

1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals

Catalog number: 157531000
100 g, Each
Thermo Scientific Chemicals

1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals

Catalog number: 157531000
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 157531000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66-71-7
IUPAC Name
1,10-phenanthroline
Molecular Formula
C12H8N2
InChI Key
DGEZNRSVGBDHLK-UHFFFAOYSA-N
SMILES
C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Appearance (Color)
White to light yellow to light pink
Infrared spectrum
Conforms
Melting point
114°C to 120°C
Titration with HClO4
>=99.0 %
Sulfated ash
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text