1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals
1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 157535000
Price (USD)/ Each
1,235.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1454.00 
Save 218.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,235.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1454.00 
Save 218.35 (15%)
Add to cart
1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals
Catalog number157535000
Price (USD)/ Each
1,235.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1454.00 
Save 218.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,10-phenanthroline
Molecular FormulaC12H8N2
InChI KeyDGEZNRSVGBDHLK-UHFFFAOYSA-N
SMILESC1=CN=C2C(C=CC3=CC=CN=C23)=C1
Molecular Weight (g/mol)180.21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light pink
Infrared spectrumConforms
Melting point114°C to 120°C
Titration with HClO4>=99.0 %
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only