4-Picoline, 99%, Thermo Scientific Chemicals
4-Picoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Picoline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 157602500
Price (USD)/ Each
33.65
Online exclusive
39.70 
Save 6.05 (15%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
33.65
Online exclusive
39.70 
Save 6.05 (15%)
Add to cart
4-Picoline, 99%, Thermo Scientific Chemicals
Catalog number157602500
Price (USD)/ Each
33.65
Online exclusive
39.70 
Save 6.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-methylpyridine
Molecular FormulaC6H7N
InChI KeyFKNQCJSGGFJEIZ-UHFFFAOYSA-N
SMILESCC1=CC=NC=C1
Molecular Weight (g/mol)93.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Infrared spectrumConforms
GC>=98.5 %
Water=<0.2 %
Refractive index1.5040 to 1.5060 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only