Thermo Scientific Chemicals

2-Piperidineethanol, 95%, Thermo Scientific Chemicals

Catalog number: 157615000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Piperidineethanol, 95%, Thermo Scientific Chemicals

Catalog number: 157615000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 157615000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1484-84-0
IUPAC Name
(2S)-2-(2-hydroxyethyl)piperidin-1-ium
Molecular Formula
C7H16NO
InChI Key
PTHDBHDZSMGHKF-ZETCQYMHSA-O
SMILES
OCC[C@@H]1CCCC[NH2+]1
Appearance (Form)
Low melting crystalline solid
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Melting point
38°C to 43°C
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text