Thermo Scientific Chemicals

Pyridine-N-oxide, 95%, Thermo Scientific Chemicals

Catalog number: 157695000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Pyridine-N-oxide, 95%, Thermo Scientific Chemicals

Catalog number: 157695000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 157695000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
694-59-7
IUPAC Name
pyridin-1-ium-1-olate
Molecular Formula
C5H5NO
InChI Key
ILVXOBCQQYKLDS-UHFFFAOYSA-N
SMILES
[O-][N+]1=CC=CC=C1
Water
=<2 % (K.F.)
Appearance (Color)
White to brown
Appearance (Form)
Crystals or crystalline solid and/or chunks
Infrared spectrum
Conforms
Melting point
61°C to 66°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text