1x1 image pixel for data collection
Pyrrole, 99%, extra pure
Pyrrole, 99%, extra pure
Pyrrole, 99%, extra pure
Pyrrole, 99%, extra pure
Thermo Scientific Chemicals

Pyrrole, 99%, extra pure

CAS: 109-97-7 | C4H5N | 67.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 157710250
Price (USD)
36.65
Special offer
Online exclusive
Ends: 26-Sep-2025
45.00
Save 8.35 (19%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 26-Sep-2025
45.00
Save 8.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS109-97-7
IUPAC Name1H-pyrrole
Molecular FormulaC4H5N
InChI KeyKAESVJOAVNADME-UHFFFAOYSA-N
SMILESN1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5070 to 1.5100 (20°C, 589 nm)
Appearance (Color)Clear colorless to brown
AppearanceMay darken on storage
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link