Thermo Scientific Chemicals

Tetraethyl orthosilicate, 98%, Thermo Scientific Chemicals

Catalog number: 157810100
10 L, Plastic Drum, Each
Thermo Scientific Chemicals

Tetraethyl orthosilicate, 98%, Thermo Scientific Chemicals

Catalog number: 157810100
10 L, Plastic Drum, Each
Quantity
Packaging
Catalog number: 157810100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-10-4
IUPAC Name
tetraethyl silicate
Molecular Formula
C8H20O4Si
InChI Key
BOTDANWDWHJENH-UHFFFAOYSA-N
SMILES
CCO[Si](OCC)(OCC)OCC
Color scale
=<20 APHA
Refractive index
1.3820 to 1.3840 (20°C, 589 nm)
Appearance (Form)
Clear liquid
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text