Thermo Scientific Chemicals

Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals

Catalog number: 157821000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals

Catalog number: 157821000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 157821000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4795-29-3
IUPAC Name
1-(oxolan-2-yl)methanamine
Molecular Formula
C5H11NO
InChI Key
YNOGYQAEJGADFJ-UHFFFAOYNA-N
SMILES
NCC1CCCO1
Refractive index
1.4540 to 1.4570 (20°C, 589 nm)
Appearance (Color)
Clear colorless to dark yellow
Appearance
May darken on storage
Titration with HClO4
>=96.0 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text