Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals
Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 157825000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4795-29-3
IUPAC Name1-(oxolan-2-yl)methanamine
Molecular FormulaC5H11NO
InChI KeyYNOGYQAEJGADFJ-UHFFFAOYNA-N
SMILESNCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4540 to 1.4570 (20°C, 589 nm)
Appearance (Color)Clear colorless to dark yellow
AppearanceMay darken on storage
Titration with HClO4>=96.0 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only