Triethylamine, 99%, pure, Thermo Scientific Chemicals
Triethylamine, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylamine, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
5 mL
100 mL
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number 157910100
Price (USD)/ Each
607.65
Online exclusive
675.00 
Save 67.35 (10%)
-
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
607.65
Online exclusive
675.00 
Save 67.35 (10%)
Add to cart
Triethylamine, 99%, pure, Thermo Scientific Chemicals
Catalog number157910100
Price (USD)/ Each
607.65
Online exclusive
675.00 
Save 67.35 (10%)
-
Add to cart
Chemical Identifiers
CAS121-44-8
IUPAC Nametriethylamine
Molecular FormulaC6H15N
InChI KeyZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILESCCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only