Trifluoroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals
Trifluoroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 157930250
Price (USD)/ Each
34.65
Online exclusive
37.80 
Save 3.15 (8%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
34.65
Online exclusive
37.80 
Save 3.15 (8%)
Add to cart
Trifluoroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals
Catalog number157930250
Price (USD)/ Each
34.65
Online exclusive
37.80 
Save 3.15 (8%)
-
Add to cart
Chemical Identifiers
CAS2923-18-4
IUPAC Namesodium trifluoroacetate
Molecular FormulaC2F3NaO2
InChI KeyUYCAUPASBSROMS-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point205°C to 207°C
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only