Thermo Scientific Chemicals

Trimethyl phosphate, 99%, Thermo Scientific Chemicals

Catalog number: 157970025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Trimethyl phosphate, 99%, Thermo Scientific Chemicals

Catalog number: 157970025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 157970025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
512-56-1
IUPAC Name
trimethyl phosphate
Molecular Formula
C3H9O4P
InChI Key
WVLBCYQITXONBZ-UHFFFAOYSA-N
SMILES
COP(=O)(OC)OC
GC
>=98.5 %
Infrared spectrum
Conforms
Water
=<0.2 %
Refractive index
1.3930 to 1.3970 (20°C, 589 nm)
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text