Trimethyl phosphate, 99%, Thermo Scientific Chemicals
Trimethyl phosphate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethyl phosphate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 157970500
Price (USD)/ Each
27.10
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
27.10
Add to cart
Trimethyl phosphate, 99%, Thermo Scientific Chemicals
Catalog number157970500
Price (USD)/ Each
27.10
-
Add to cart
Chemical Identifiers
CAS512-56-1
IUPAC Nametrimethyl phosphate
Molecular FormulaC3H9O4P
InChI KeyWVLBCYQITXONBZ-UHFFFAOYSA-N
SMILESCOP(=O)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Water=<0.2 %
Refractive index1.3930 to 1.3970 (20°C, 589 nm)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only