Thermo Scientific Chemicals

Tryptamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 157980050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tryptamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 157980050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 157980050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
343-94-2
IUPAC Name
hydrogen 2-(1H-indol-3-yl)ethan-1-amine chloride
Molecular Formula
C10H13ClN2
InChI Key
KDFBGNBTTMPNIG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCC1=CNC2=CC=CC=C12
Appearance (Form)
Powder
Appearance (Color)
Beige to light orange
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Sulfated ash
=<0.2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text