Tryptamine hydrochloride, 98%, Thermo Scientific Chemicals
Tryptamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tryptamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 157980100
Price (USD)/ Each
63.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.70 
Save 11.05 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
63.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.70 
Save 11.05 (15%)
Add to cart
Tryptamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog number157980100
Price (USD)/ Each
63.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.70 
Save 11.05 (15%)
-
Add to cart
Chemical Identifiers
CAS343-94-2
IUPAC Namehydrogen 2-(1H-indol-3-yl)ethan-1-amine chloride
Molecular FormulaC10H13ClN2
InChI KeyKDFBGNBTTMPNIG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Beige to light orange
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Sulfated ash=<0.2 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only